Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CAMK4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001323374.1 | NP_001310303.1 | 12 | 1552 | ||
NM_001323375.1 | NP_001310304.1 | 12 | 1584 | |||
NM_001323376.1 | NP_001310305.1 | 9 | 1554 | |||
NM_001323377.1 | NP_001310306.1 | 10 | 1312 | |||
NM_001744.5 | NP_001735.1 | 11 | 1700 | |||
GenBank mRNA | AK311662.1 | - | 9 | 1409 | ||
AL833368.1 | - | 2 | 1462 | |||
BC016695.1 | - | 11 | 1544 | |||
BC025687.1 | - | 11 | 1542 | 5205478 | ||
BX374853.2 | - | 1 | 454 | |||
D30742.1 | - | 11 | 1530 | |||
L17000.1 | - | 11 | 1494 | |||
L24959.1 | - | 12 | 1552 |