Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CAMLG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001745.3 | NP_001736.1 | 2 | 641 | ||
XM_011543653.1 | XP_011541955.1 | 2 | 1194 | |||
GenBank mRNA | AF068179.1 | - | 2 | 358 | ||
AK313156.1 | - | 2 | 565 | |||
BC130325.1 | - | 2 | 573 | 40146356 | ||
BG720084.1 | - | 2 | 613 | |||
U18242.1 | - | 2 | 536 |