Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CANX | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024649.1 | NP_001019820.1 | 15 | 2034 | ||
NM_001746.3 | NP_001737.1 | 15 | 2087 | |||
XM_011534664.1 | XP_011532966.1 | 15 | 2452 | |||
XM_011534665.2 | XP_011532967.1 | 15 | 2060 | |||
GenBank mRNA | AK129990.1 | - | 9 | 1050 | ||
AK130080.1 | - | 5 | 569 | |||
AK294702.1 | - | 15 | 2090 | |||
BC003552.1 | - | 15 | 2021 | 3546389 | ||
BC042843.1 | - | 15 | 1968 | 4342723 | ||
L10284.1 | - | 15 | 1999 | |||
L18887.1 | - | 15 | 2005 | |||
M94859.1 | - | 15 | 2005 | |||
M98452.1 | - | 8 | 1202 |