Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other CAST SIRNA Assays ›
Gene Symbol:
CAST
Gene Name:
calpastatin
Gene Aliases:
BS-17, PLACK
Chromosome Location:
Chr.5: 96662037 - 96774683 on Build GRCh38
UniGene ID:
Hs.436186
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
145948
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: CAST
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001042440.3
NP_001035905.1
14
1324
NM_001190442.1
NP_001177371.1
15
1196
NM_001284212.2
NP_001271141.1
12
975
NM_001284213.2
NP_001271142.1
11
882
NM_173060.3
NP_775083.1
13
1032
XM_006714696.3
XP_006714759.1
17
1338
XM_006714697.3
XP_006714760.1
16
1355
XM_006714698.3
XP_006714761.1
15
1316
XM_006714699.3
XP_006714762.1
16
1279
XM_006714700.3
XP_006714763.1
15
1298
XM_006714701.3
XP_006714764.1
15
1314
XM_006714702.3
XP_006714765.1
15
1190
XM_006714703.3
XP_006714766.1
14
1275
XM_006714704.3
XP_006714767.1
15
1223
XM_006714705.3
XP_006714768.1
14
1232
XM_006714706.3
XP_006714769.1
14
1150
XM_006714707.3
XP_006714770.1
14
1101
XM_006714708.3
XP_006714771.1
13
1061
XM_006714709.3
XP_006714772.1
13
1024
XM_006714710.3
XP_006714773.1
12
979
XM_006714711.3
XP_006714774.1
12
1024
XM_006714712.3
XP_006714775.1
11
971
XM_006714715.3
XP_006714778.1
2
914
XM_011543654.2
XP_011541956.1
15
1274
XM_011543655.2
XP_011541957.1
16
1355
XM_011543656.2
XP_011541958.1
14
1218
XM_011543657.2
XP_011541959.1
14
1100
XM_011543658.2
XP_011541960.1
12
953
XM_017009911.1
XP_016865400.1
15
1316
XM_017009912.1
XP_016865401.1
16
1327
XM_017009913.1
XP_016865402.1
16
1296
XM_017009914.1
XP_016865403.1
15
1298
XM_017009915.1
XP_016865404.1
16
1300
XM_017009916.1
XP_016865405.1
15
1314
XM_017009917.1
XP_016865406.1
15
1232
XM_017009918.1
XP_016865407.1
14
1259
XM_017009919.1
XP_016865408.1
15
1316
XM_017009920.1
XP_016865409.1
14
1142
XM_017009921.1
XP_016865410.1
14
1159
XM_017009922.1
XP_016865411.1
13
1193
XM_017009923.1
XP_016865412.1
13
1193
XM_017009924.1
XP_016865413.1
14
1259
XM_017009925.1
XP_016865414.1
13
1111
XM_017009926.1
XP_016865415.1
13
1111
XM_017009927.1
XP_016865416.1
14
1100
XM_017009928.1
XP_016865417.1
13
1099
XM_017009929.1
XP_016865418.1
13
1018
XM_017009930.1
XP_016865419.1
12
985
XM_017009931.1
XP_016865420.1
12
979
XM_017009932.1
XP_016865421.1
11
1042
GenBank mRNA
AB208816.1
-
15
1160
AF327443.1
-
10
874
AK296919.1
-
15
1154
AK297879.1
-
13
974
AK298541.1
-
14
1101
AK299384.1
-
14
1005
AK299402.1
-
14
1081
AK300624.1
-
11
865
AK302206.1
-
2
188
AK303836.1
-
7
555
AK303900.1
-
10
768
AK311585.1
-
2
366
AK311598.1
-
2
242
AK314290.1
-
12
977
AK315146.1
-
12
976
AL832174.1
-
13
1017
AL832349.1
-
13
1020
AY189741.1
-
3
187
BC013579.1
-
14
1027
3878564
BC016066.1
-
14
1296
BC078149.1
-
9
610
BT009783.1
-
11
809
BX648397.1
-
12
926
D16217.1
-
14
1094
U26724.2
-
5
457
U31345.1
-
3
204
Back To Top
More Information
Panther Classification:
Molecular Function -
protein-binding activity modulator
protease inhibitor
Gene Ontology Categories:
Function(s)
Process(es)
liver development
egg activation
brain development
myoblast fusion
aging
negative regulation of endopeptidase activity
organ regeneration
myoblast differentiation
negative regulation of cell cycle arrest
cell-cell adhesion
negative regulation of type B pancreatic cell apoptotic process
protease binding
endopeptidase inhibitor activity
protein binding
calcium-dependent cysteine-type endopeptidase inhibitor activity
poly(A) RNA binding
cadherin binding involved in cell-cell adhesion
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-cccc65c97-qtpg5:80/10.239.161.70:80.
git-commit: 2b810224bbfb944e11adb8d8c7a69f36ca15064f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.1-offline