Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CNN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
RefSeq | NM_001303499.1 | NP_001290428.1 | 2 | 551 | ||
NM_001303501.1 | NP_001290430.1 | 2 | 551 | |||
NM_004368.3 | NP_004359.1 | 2 | 551 | |||
NM_201277.2 | NP_958434.1 | 2 | 551 | |||
GenBank mRNA | AK024835.1 | - | 2 | 197 | ||
AK126391.1 | - | 2 | 216 | |||
AK222924.1 | - | 2 | 187 | |||
AK293171.1 | - | 2 | 197 | |||
AK297774.1 | - | 2, 3 | 136 | |||
AK300788.1 | - | 2 | 186 | |||
AY927514.1 | - | LIT3524 AY927514 |
2 | 135 | ||
BC141818.1 | - | 2 | 194 | 40148129 | ||
BC141833.1 | - | 2 | 194 | 40148834 | ||
BC148264.1 | - | 2 | 186 | 8860204 | ||
BQ876824.1 | - | 2 | 126 | |||
BQ919304.1 | - | 2 | 527 | |||
CR541810.1 | - | 2 | 164 | |||
CR541831.1 | - | 2 | 164 | |||
D83735.1 | - | 2 | 191 |