Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CD34 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025109.1 | NP_001020280.1 | 4, 5 | 859 | ||
NM_001773.2 | NP_001764.1 | 4, 5 | 859 | |||
GenBank mRNA | AF202879.1 | - | 2, 3 | 104 | ||
AF523361.1 | - | 4, 5 | 672 | |||
AF538601.1 | - | 3, 4 | 510 | |||
AK291529.1 | - | 4, 5 | 612 | |||
AK293030.1 | - | 4, 5 | 636 | |||
AK294381.1 | - | 3, 4 | 383 | |||
AK300835.1 | - | 5, 6 | 676 | |||
BC039146.1 | - | 4, 5 | 609 | 4746591 | ||
M81104.1 | - | 4, 5 | 858 | |||
S53910.1 | - | 4, 5 | 691 | |||
S53911.1 | - | 4, 5 | 691 |