Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CD34 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025109.1 | NP_001020280.1 | 8 | 2520 | ||
NM_001773.2 | NP_001764.1 | 8 | 2715 | |||
GenBank mRNA | AK091890.1 | - | 5 | 2237 | ||
BC039146.1 | - | 8 | 2270 | 4746591 | ||
BX538076.1 | - | 5 | 2448 | |||
M81104.1 | - | 8 | 2520 | |||
S53910.1 | - | 8 | 2350 | |||
S53911.1 | - | 8 | 2544 |