Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CD37 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040031.1 | NP_001035120.1 | 6 | 655 | ||
NM_001774.2 | NP_001765.1 | 6 | 689 | |||
XM_005259435.3 | XP_005259492.1 | 6 | 691 | |||
XM_005259436.3 | XP_005259493.1 | 6 | 687 | |||
XM_011527542.2 | XP_011525844.1 | 6 | 652 | |||
XM_011527543.2 | XP_011525845.1 | 6 | 757 | |||
XM_011527544.2 | XP_011525846.1 | 6 | 726 | |||
XM_017027513.1 | XP_016883002.1 | 6 | 687 | |||
GenBank mRNA | AK058093.1 | - | 7 | 1036 | ||
AK301016.1 | - | 6 | 679 | |||
AK301328.1 | - | 6 | 682 | |||
AK316348.1 | - | 6 | 579 | |||
AL833693.1 | - | 5 | 1733 | |||
BC106752.1 | - | 6 | 606 | 40023348 | ||
BC110379.1 | - | 6 | 641 | 5752290 | ||
BT019851.1 | - | 6 | 568 | |||
CR541877.1 | - | 6 | 568 | |||
X14046.1 | - | 6 | 631 |