Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COL6A3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004369.3 | NP_004360.2 | 5 | 1730 | ||
NM_057164.4 | NP_476505.3 | 3 | 509 | |||
NM_057165.4 | NP_476506.3 | 4 | 1112 | |||
NM_057166.4 | NP_476507.3 | 3 | 509 | |||
NM_057167.3 | NP_476508.2 | 4 | 1112 | |||
XM_005246065.1 | XP_005246122.1 | 5 | 1687 | |||
XM_005246066.1 | XP_005246123.1 | 2 | 233 | |||
XM_006712253.1 | XP_006712316.1 | 5 | 1687 | |||
XM_011510574.1 | XP_011508876.1 | 5 | 1687 | |||
XM_017003303.1 | XP_016858792.1 | 10 | 1737 | |||
GenBank mRNA | AK092021.1 | - | 3 | 435 | ||
AK303290.1 | - | 3 | 476 | |||
AK304870.1 | - | 4 | 1047 | |||
AK308451.1 | - | 5 | 1687 | |||
BC144595.1 | - | 3 | 450 | 9053125 | ||
BC150625.1 | - | 3 | 450 | 9056995 | ||
BC171790.1 | - | 3 | 450 | 9054444 | ||
BX647500.1 | - | 4 | 1074 | |||
DC411101.1 | - | 3 | 509 | |||
X52022.1 | - | 5 | 1700 |