Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CDH2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001308176.1 | NP_001295105.1 | 2 | 324 | ||
NM_001792.4 | NP_001783.2 | 3 | 804 | |||
XM_011525788.1 | XP_011524090.1 | 3 | 469 | |||
XM_017025514.1 | XP_016881003.1 | 3 | 775 | |||
GenBank mRNA | AK302831.1 | - | 2 | 314 | ||
BC036470.1 | - | 3 | 496 | 5264944 | ||
CN263272.1 | - | 3 | 512 | |||
DC403359.1 | - | 2 | 324 | |||
S42303.1 | - | 3 | 838 | |||
X54315.1 | - | 3 | 481 |