Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CDH2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001308176.1 | NP_001295105.1 | 9 | 1518 | ||
NM_001792.4 | NP_001783.2 | 10 | 1998 | |||
XM_011525788.1 | XP_011524090.1 | 10 | 1663 | |||
XM_017025514.1 | XP_016881003.1 | 10 | 1969 | |||
GenBank mRNA | AK302831.1 | - | 9 | 1508 | ||
BC036470.1 | - | 10 | 1690 | 5264944 | ||
M34064.1 | - | 7 | 1097 | |||
S42303.1 | - | 10 | 2032 | |||
X54315.1 | - | 10 | 1675 | |||
X57548.1 | - | 7 | 1097 |