Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CDH2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001308176.1 | NP_001295105.1 | 8, 9 | 1297 | ||
NM_001792.4 | NP_001783.2 | 9, 10 | 1777 | |||
XM_011525788.1 | XP_011524090.1 | 9, 10 | 1442 | |||
XM_017025514.1 | XP_016881003.1 | 9, 10 | 1748 | |||
GenBank mRNA | AK302831.1 | - | 8, 9 | 1287 | ||
M34064.1 | - | 6, 7 | 876 | |||
S42303.1 | - | 9, 10 | 1811 | |||
X54315.1 | - | 9, 10 | 1454 | |||
X57548.1 | - | 6, 7 | 876 |