Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EPHA5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001281765.2 | NP_001268694.1 | 3 | 1125 | ||
NM_001281766.2 | NP_001268695.1 | 3 | 1125 | |||
NM_001281767.2 | NP_001268696.1 | 3 | 1125 | |||
NM_001318761.1 | NP_001305690.1 | 3 | 243 | |||
NM_004439.7 | NP_004430.4 | 3 | 1125 | |||
NM_182472.4 | NP_872272.2 | 3 | 1125 | |||
XM_005265653.3 | XP_005265710.1 | 3 | 1468 | |||
XM_011531735.2 | XP_011530037.1 | 3 | 1468 | |||
XM_017007878.1 | XP_016863367.1 | 3 | 1468 | |||
XM_017007879.1 | XP_016863368.1 | 3 | 1468 | |||
XM_017007880.1 | XP_016863369.1 | 3 | 1468 | |||
XM_017007881.1 | XP_016863370.1 | 3 | 1468 | |||
GenBank mRNA | AB209375.1 | - | 3 | 1125 | ||
AK307826.1 | - | 3 | 564 | |||
BC136257.1 | - | 3 | 392 | 9020244 | ||
BC136258.1 | - | 3 | 392 | 9020245 | ||
BC143427.1 | - | 3 | 392 | 9051935 | ||
BX537946.1 | - | 3 | 244 | |||
L36644.1 | - | 3 | 300 |