Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CKM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001824.4 | NP_001815.2 | 8 | 1479 | ||
XM_017026240.1 | XP_016881729.1 | 7 | 1364 | |||
GenBank mRNA | AJ572902.1 | - | 1 | 241 | ||
AK129878.1 | - | 8 | 1377 | |||
AK300730.1 | - | 8 | 1267 | |||
BC007462.1 | - | 8 | 1377 | 3934232 | ||
M14780.1 | - | 8 | 1382 |