Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CKMT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001099735.1 | NP_001093205.1 | 2 | 215 | ||
NM_001099736.1 | NP_001093206.1 | 2 | 219 | |||
NM_001825.2 | NP_001816.2 | 3 | 379 | |||
GenBank mRNA | AK122769.1 | - | 2 | 198 | ||
AK313293.1 | - | 2 | 206 | |||
BC029140.1 | - | 2 | 196 | 5195322 | ||
CR450315.1 | - | 1 | 137 | |||
DA826840.1 | - | 3 | 375 | |||
DB479765.1 | - | 2 | 250 | |||
DC308656.1 | - | 2 | 219 | |||
J05401.1 | - | 2 | 335 |