Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CNTN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256063.1 | NP_001242992.1 | 5 | 617 | ||
NM_001256064.1 | NP_001242993.1 | 5 | 830 | |||
NM_001843.3 | NP_001834.2 | 5 | 617 | |||
NM_175038.2 | NP_778203.1 | 3 | 433 | |||
XM_005268651.2 | XP_005268708.1 | 5 | 980 | |||
XM_006719241.2 | XP_006719304.1 | 5 | 604 | |||
XM_011537926.2 | XP_011536228.1 | 5 | 569 | |||
XM_011537927.2 | XP_011536229.1 | 5 | 626 | |||
XM_017018821.1 | XP_016874310.1 | 5 | 620 | |||
XM_017018822.1 | XP_016874311.1 | 5 | 569 | |||
XM_017018823.1 | XP_016874312.1 | 5 | 980 | |||
XM_017018824.1 | XP_016874313.1 | 5 | 604 | |||
XM_017018825.1 | XP_016874314.1 | 5 | 569 | |||
XM_017018826.1 | XP_016874315.1 | 5 | 604 | |||
XM_017018827.1 | XP_016874316.1 | 5 | 626 | |||
GenBank mRNA | AK289544.1 | - | 5 | 709 | ||
AK289698.1 | - | 5 | 559 | |||
BC036569.2 | - | 5 | 556 | 5273941 | ||
U07819.1 | - | 4 | 390 | |||
U07820.1 | - | 3 | 438 | |||
Z21488.1 | - | 5 | 511 |