Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NCAN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004386.2 | NP_004377.2 | 3 | 398 | ||
GenBank mRNA | AB210004.1 | - | 3 | 342 | ||
AF026547.1 | - | 2 | 300 | |||
CN282070.1 | - | 3 | 398 |