Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COL6A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001848.2 | NP_001839.2 | 3 | 496 | ||
GenBank mRNA | AK298968.1 | - | 3 | 475 | ||
AK299396.1 | - | 3 | 478 | |||
AK307989.1 | - | 3 | 475 | |||
BC052575.1 | - | 3 | 476 | 6598940 | ||
CN369026.1 | - | 3 | 494 | |||
X15879.1 | - | 3 | 442 |