Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COL6A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001848.2 | NP_001839.2 | 35 | 4061 | ||
GenBank mRNA | AK299396.1 | - | 27 | 2001 | ||
BC005159.2 | - | 9 | 2211 | |||
BC022236.1 | - | 6 | 2180 | |||
BM686857.1 | - | 1 | 469 | |||
BM707144.1 | - | 1 | 311 | |||
X15880.1 | - | 9 | 2128 |