Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COL8A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001850.4 | NP_001841.2 | 4 | 403 | ||
NM_020351.3 | NP_065084.2 | 3 | 341 | |||
GenBank mRNA | AY550974.1 | - | 2 | 201 | ||
BC013581.1 | - | 4 | 381 | 3875911 | ||
BT009917.1 | - | 1 | 144 | |||
CA393523.1 | - | 3 | 327 | |||
CN484310.1 | - | 3 | 327 | |||
X57527.1 | - | 1 | 144 |