Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COL8A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001850.4 | NP_001841.2 | 5 | 2286 | ||
NM_020351.3 | NP_065084.2 | 4 | 2224 | |||
GenBank mRNA | AY550974.1 | - | 3 | 2087 | ||
BC013581.1 | - | 5 | 2264 | 3875911 | ||
BG938623.1 | - | 1 | 98 | |||
BT009917.1 | - | 2 | 2027 | |||
X57527.1 | - | 2 | 2027 |