Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COL9A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001851.4 | NP_001842.3 | 38 | 3101 | ||
NM_078485.3 | NP_511040.2 | 32 | 2369 | |||
XM_011535429.2 | XP_011533731.1 | 39 | 3131 | |||
XM_011535430.2 | XP_011533732.1 | 33 | 2397 | |||
XM_017010246.1 | XP_016865735.1 | 36 | 2488 | |||
XM_017010247.1 | XP_016865736.1 | 30 | 2344 | |||
GenBank mRNA | AK097582.1 | - | 10 | 1647 | ||
BC063646.1 | - | 32 | 2343 | 4914907 |