Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COL16A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001856.3 | NP_001847.3 | 5 | 720 | ||
XM_005270481.1 | XP_005270538.1 | 5 | 715 | |||
XM_011540722.1 | XP_011539024.1 | 5 | 715 | |||
XM_011540723.1 | XP_011539025.1 | 5 | 715 | |||
XM_011540724.1 | XP_011539026.1 | 5 | 715 | |||
XM_011540726.2 | XP_011539028.1 | 2 | 231 | |||
XM_011540729.1 | XP_011539031.1 | 5 | 715 | |||
XM_011540730.1 | XP_011539032.1 | 5 | 715 | |||
XM_017000338.1 | XP_016855827.1 | 5 | 715 | |||
XM_017000339.1 | XP_016855828.1 | 5 | 715 | |||
GenBank mRNA | AB209571.1 | - | 1 | 27 | ||
M92642.1 | - | 6 | 508 |