Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COL16A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001856.3 | NP_001847.3 | 71 | 5241 | ||
XM_005270481.1 | XP_005270538.1 | 70 | 5188 | |||
XM_011540722.1 | XP_011539024.1 | 70 | 5191 | |||
XM_011540723.1 | XP_011539025.1 | 69 | 5143 | |||
XM_011540724.1 | XP_011539026.1 | 70 | 5113 | |||
XM_011540726.2 | XP_011539028.1 | 66 | 4659 | |||
XM_011540727.2 | XP_011539029.1 | 61 | 3997 | |||
XM_011540728.2 | XP_011539030.1 | 49 | 3479 | |||
XM_017000338.1 | XP_016855827.1 | 70 | 5203 | |||
XM_017000339.1 | XP_016855828.1 | 70 | 5200 | |||
XM_017000340.1 | XP_016855829.1 | 48 | 3442 | |||
XM_017000341.1 | XP_016855830.1 | 48 | 3232 | |||
GenBank mRNA | AB209571.1 | - | 65 | 4455 | ||
M92642.1 | - | 72 | 5026 | |||
S57132.1 | - | 54 | 3622 |