Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CRHBP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001882.3 | NP_001873.2 | 7 | 1562 | ||
GenBank mRNA | AK223457.1 | - | 7 | 1439 | ||
BC018038.1 | - | 7 | 1372 | 4812893 |