Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CRYZ | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001130042.1 | NP_001123514.1 | 3, 4 | 618 | ||
NM_001130043.1 | NP_001123515.1 | 2, 3 | 579 | |||
NM_001889.3 | NP_001880.2 | 2, 3 | 579 | |||
XM_005270491.4 | XP_005270548.1 | 3, 4 | 256 | |||
XM_011540747.1 | XP_011539049.1 | 2, 3 | 198 | |||
XM_017000367.1 | XP_016855856.1 | 3, 4 | 253 | |||
XM_017000368.1 | XP_016855857.1 | 2, 3 | 266 | |||
GenBank mRNA | AB209714.1 | - | 3, 4 | 250 | ||
AK128794.1 | - | 2, 3 | 184 | |||
AK223150.1 | - | 2, 3 | 211 | |||
AK223201.1 | - | 2, 3 | 211 | |||
AK314813.1 | - | 2, 3 | 219 | |||
BC039578.1 | - | 2, 3 | 201 | 6067125 | ||
BC070058.1 | - | 3, 4 | 260 | 5267616 | ||
DA256305.1 | - | 2, 3 | 263 | |||
L13278.1 | - | 1, 2 | 123 |