Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DFNA5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127453.1 | NP_001120925.1 | 3 | 452 | ||
NM_001127454.1 | NP_001120926.1 | 2 | 467 | |||
NM_004403.2 | NP_004394.1 | 3 | 697 | |||
XM_017011802.1 | XP_016867291.1 | 2 | 223 | |||
GenBank mRNA | AF007790.2 | - | 2 | 352 | ||
AF073308.1 | - | 3 | 421 | |||
AK094714.1 | - | 5 | 1025 | |||
AK314402.1 | - | 3 | 437 | |||
BC099911.1 | - | 3 | 413 | 5287362 | ||
BC125065.1 | - | 3 | 419 | 40117291 | ||
BC125066.1 | - | 2 | 189 | |||
DA820184.1 | - | 3 | 452 |