Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CTGF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001901.2 | NP_001892.1 | 5 | 1622 | ||
GenBank mRNA | AF127918.1 | - | 1 | 350 | ||
AK092280.1 | - | 5 | 1473 | |||
AK125220.1 | - | 5 | 1404 | |||
AY395801.1 | - | 5 | 1526 | |||
AY550024.1 | - | 5 | 1516 | |||
BC087839.1 | - | 5 | 1598 | 6181580 | ||
M92934.1 | - | 5 | 1542 | |||
U14750.1 | - | 4 | 1196 | |||
X78947.1 | - | 5 | 1560 |