Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CTNNA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290307.2 | NP_001277236.1 | 7 | 1128 | ||
NM_001290309.2 | NP_001277238.1 | 6 | 932 | |||
NM_001290310.2 | NP_001277239.1 | 7 | 965 | |||
NM_001323982.1 | NP_001310911.1 | 8 | 1598 | |||
NM_001323983.1 | NP_001310912.1 | 7 | 1280 | |||
NM_001323984.1 | NP_001310913.1 | 8 | 1146 | |||
NM_001323985.1 | NP_001310914.1 | 7 | 1128 | |||
NM_001323986.1 | NP_001310915.1 | 7 | 1128 | |||
NM_001903.4 | NP_001894.2 | 7 | 1128 | |||
GenBank mRNA | AK094151.1 | - | 4 | 337 | ||
AK296708.1 | - | 7 | 936 | |||
AK300439.1 | - | 6 | 899 | |||
AK304267.1 | - | 7 | 1023 | |||
BC000385.2 | - | 7 | 1066 | 2821111 | ||
D13866.1 | - | 7 | 1067 | |||
D14705.1 | - | 7 | 1092 | |||
L23805.1 | - | 6 | 1005 | |||
U03100.1 | - | 6 | 1005 |