Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CTSB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001908.4 | NP_001899.1 | 5, 6 | 627 | ||
NM_147780.3 | NP_680090.1 | 7, 8 | 789 | |||
NM_147781.3 | NP_680091.1 | 6, 7 | 746 | |||
NM_147782.3 | NP_680092.1 | 6, 7 | 715 | |||
NM_147783.3 | NP_680093.1 | 6, 7 | 701 | |||
XM_006716244.2 | XP_006716307.1 | 7, 8 | 813 | |||
XM_006716245.2 | XP_006716308.1 | 5, 6 | 917 | |||
XM_011543812.2 | XP_011542114.1 | 7, 8 | 722 | |||
XM_017013097.1 | XP_016868586.1 | 6, 7 | 703 | |||
XM_017013098.1 | XP_016868587.1 | 6, 7 | 703 | |||
XM_017013099.1 | XP_016868588.1 | 7, 8 | 745 | |||
XM_017013100.1 | XP_016868589.1 | 6, 7 | 647 | |||
XM_017013101.1 | XP_016868590.1 | 2, 3 | 170 | |||
GenBank mRNA | AK075393.1 | - | 5, 6 | 603 | ||
AK092070.1 | - | 6, 7 | 638 | |||
AK130184.1 | - | 5, 6 | 564 | |||
AK290239.1 | - | 5, 6 | 564 | |||
AK297687.1 | - | 3, 4 | 328 | |||
AL543654.3 | - | 6, 7 | 675 | |||
BC010240.1 | - | 6, 7 | 644 | 4154307 | ||
BC095408.1 | - | 5, 6 | 564 | 30334082 | ||
BX647952.1 | - | 5, 6 | 567 | |||
BX648239.1 | - | 2, 3 | 4638 | |||
L16510.1 | - | 6, 7 | 629 | |||
M13230.1 | - | 1, 2 | 62 | |||
M14221.1 | - | 6, 7 | 646 | |||
Y18460.1 | - | 1, 2 | 134 |