Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CTSE | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001317331.1 | NP_001304260.1 | 7 | 1478 | ||
NM_001910.3 | NP_001901.1 | 9 | 1777 | |||
NM_148964.2 | NP_683865.1 | 8 | 1635 | |||
XM_011509244.1 | XP_011507546.1 | 9 | 1782 | |||
XM_011509245.1 | XP_011507547.1 | 8 | 1640 | |||
GenBank mRNA | AJ250716.1 | - | 8 | 1621 | ||
AJ250717.1 | - | 9 | 1763 | |||
BC042537.1 | - | 9 | 1744 | 5174814 | ||
J05036.1 | - | 9 | 1708 |