Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DEFA6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001926.3 | NP_001917.1 | 2 | 252 | ||
GenBank mRNA | BC069667.1 | - | 2 | 252 | 7262522 | |
BC069710.1 | - | 2 | 252 | 7262534 | ||
BC069728.1 | - | 2 | 252 | 7262546 | ||
BC069769.1 | - | 2 | 254 | 7262558 | ||
BC093951.1 | - | 2 | 232 | 7939796 | ||
BC093953.1 | - | 2 | 232 | 7939798 | ||
M98331.1 | - | 2 | 229 |