Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other TRDMT1 SIRNA Assays ›
Gene Symbol:
TRDMT1
Gene Name:
tRNA aspartic acid methyltransferase 1
Gene Aliases:
DMNT2, DNMT2, MHSAIIP, PUMET, RNMT1
Chromosome Location:
Chr.10: 17136785 - 17202071 on Build GRCh38
UniGene ID:
Hs.351665
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
14618
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: TRDMT1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001321006.1
NP_001307935.1
6
1021
NM_001321007.1
NP_001307936.1
9
1394
NM_004412.6
NP_004403.1
8
1164
XM_005252372.4
XP_005252429.1
8
1159
XM_005252374.4
XP_005252431.1
8
1159
XM_005252375.4
XP_005252432.1
6
1021
XM_005252378.3
XP_005252435.1
9
990
XM_006717388.3
XP_006717451.1
8
1159
XM_011519343.2
XP_011517645.1
7
1093
XM_011519344.2
XP_011517646.1
7
1082
XM_011519345.2
XP_011517647.1
7
1495
XM_017015800.1
XP_016871289.1
6
1021
XM_017015801.1
XP_016871290.1
6
1021
XM_017015802.1
XP_016871291.1
8
3378
XM_017015803.1
XP_016871292.1
9
992
XM_017015804.1
XP_016871293.1
9
830
XM_017015805.1
XP_016871294.1
8
1027
XM_017015806.1
XP_016871295.1
9
990
XM_017015807.1
XP_016871296.1
9
1011
XM_017015808.1
XP_016871297.1
9
825
XM_017015809.1
XP_016871298.1
5
944
XM_017015810.1
XP_016871299.1
8
924
XM_017015811.1
XP_016871300.1
6
890
XM_017015812.1
XP_016871301.1
6
684
GenBank mRNA
AF012128.1
-
8
734
AF045888.1
-
8
733
AJ223333.1
-
8
775
AK291417.1
-
8
733
AK294100.1
-
7
663
AK299027.1
-
6
801
AK303428.1
-
3
381
BC047733.1
-
8
734
6473993
BX537961.1
-
9
984
CR450316.1
-
8
727
HQ637546.1
-
6
583
HQ637547.1
-
7
650
HQ637548.1
-
6
584
HQ637549.1
-
7
661
Back To Top
More Information
Panther Classification:
Molecular Function -
RNA metabolism protein
RNA processing factor
nucleic acid metabolism protein
Gene Ontology Categories:
Function(s)
Process(es)
response to amphetamine
tRNA modification
tRNA methylation
C-5 methylation of cytosine
RNA binding
DNA (cytosine-5-)-methyltransferase activity
tRNA methyltransferase activity
tRNA (cytosine-5-)-methyltransferase activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-cccc65c97-hnnbr:80/10.239.164.27:80.
git-commit: 2b810224bbfb944e11adb8d8c7a69f36ca15064f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.1-offline