Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DPEP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001128141.2 | NP_001121613.1 | 7, 8 | 972 | ||
NM_004413.3 | NP_004404.1 | 7, 8 | 1060 | |||
XM_005256285.4 | XP_005256342.1 | 7, 8 | 1010 | |||
XM_005256286.3 | XP_005256343.1 | 7, 8 | 922 | |||
XM_011522925.2 | XP_011521227.1 | 7, 8 | 1020 | |||
XM_017023009.1 | XP_016878498.1 | 7, 8 | 1083 | |||
GenBank mRNA | AK309577.1 | - | 5, 6 | 1104 | ||
BC017023.1 | - | 6, 7 | 891 | 3846046 | ||
BT006664.1 | - | 6, 7 | 769 | |||
CR457404.1 | - | 6, 7 | 769 | |||
D13138.1 | - | 6, 7 | 812 | |||
J05257.1 | - | 7, 8 | 1064 |