Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EFNA5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001962.2 | NP_001953.1 | 4, 5 | 852 | ||
XM_011543250.2 | XP_011541552.1 | 4, 5 | 665 | |||
XM_011543251.2 | XP_011541553.1 | 4, 5 | 2178 | |||
XM_017009205.1 | XP_016864694.1 | 4, 5 | 1941 | |||
GenBank mRNA | BC075054.2 | - | 4, 5 | 619 | 30915365 | |
BC075055.2 | - | 4, 5 | 619 | 30915581 | ||
U26403.1 | - | 4, 5 | 852 |