Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EPS15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159969.1 | NP_001153441.1 | 3 | 522 | ||
NM_001981.2 | NP_001972.1 | 15 | 1391 | |||
XM_005270618.2 | XP_005270675.1 | 14 | 1332 | |||
XM_017000615.1 | XP_016856104.1 | 15 | 1533 | |||
XM_017000616.1 | XP_016856105.1 | 15 | 1533 | |||
XM_017000617.1 | XP_016856106.1 | 15 | 1422 | |||
XM_017000618.1 | XP_016856107.1 | 14 | 1332 | |||
XM_017000619.1 | XP_016856108.1 | 16 | 1415 | |||
XM_017000620.1 | XP_016856109.1 | 15 | 1582 | |||
GenBank mRNA | AK294297.1 | - | 4 | 371 | ||
AK308128.1 | - | 3 | 486 | |||
AK313396.1 | - | 15 | 1340 | |||
BX647676.1 | - | 3 | 506 | |||
DA801048.1 | - | 3 | 522 | |||
DQ367924.1 | - | 15 | 1299 | |||
U07707.1 | - | 15 | 1320 | |||
Z29064.1 | - | 15 | 1391 |