Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PHC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004426.2 | NP_004417.2 | 2 | 776 | ||
XM_005253334.2 | XP_005253391.1 | 6, 7 | 776 | |||
XM_011520599.2 | XP_011518901.1 | 6, 7 | 779 | |||
XM_011520600.2 | XP_011518902.1 | 5, 6 | 653 | |||
XM_011520603.2 | XP_011518905.1 | 6, 7 | 883 | |||
XM_017018955.1 | XP_016874444.1 | 6, 7 | 1171 | |||
XM_017018956.1 | XP_016874445.1 | 5, 6 | 629 | |||
XM_017018957.1 | XP_016874446.1 | 5, 6 | 602 | |||
XM_017018958.1 | XP_016874447.1 | 5, 6 | 578 | |||
GenBank mRNA | AB209063.1 | - | 7, 8 | 952 | ||
AK293893.1 | - | 6, 7 | 800 | |||
BC073964.1 | - | 3 | 630 | 5788132 | ||
BC157850.1 | - | 2 | 682 | 9057069 | ||
BX640639.1 | - | 7, 8 | 902 | |||
CN388316.1 | - | 1 | 176 | |||
U89277.1 | - | 6, 7 | 755 |