Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: F13B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001994.2 | NP_001985.2 | 9 | 1516 | ||
XM_011509283.2 | XP_011507585.1 | 9 | 1513 | |||
XM_011509284.2 | XP_011507586.1 | 9 | 1510 | |||
XM_011509286.2 | XP_011507588.1 | 9 | 1524 | |||
GenBank mRNA | AK290560.1 | - | 9 | 1513 | ||
M14057.1 | - | 9 | 1470 |