Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EFNA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004428.2 | NP_004419.2 | 2 | 218 | ||
NM_182685.1 | NP_872626.1 | 2 | 218 | |||
GenBank mRNA | AK057845.1 | - | 2 | 631 | ||
AK289589.1 | - | 2 | 215 | |||
AK308965.1 | - | 2 | 215 | |||
AK313117.1 | - | 2 | 262 | |||
AL833641.1 | - | 2 | 216 | |||
BC032698.1 | - | 2 | 211 | 5402648 | ||
BC095432.1 | - | 2 | 200 | 30376031 | ||
CR457416.1 | - | 2 | 113 | |||
M57730.1 | - | 2 | 186 |