Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CLK2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001294338.1 | NP_001281267.1 | 6 | 967 | ||
NM_001294339.1 | NP_001281268.1 | 5 | 879 | |||
NM_003993.3 | NP_003984.2 | 6 | 964 | |||
XM_005244878.1 | XP_005244935.1 | 6 | 946 | |||
XM_011509143.1 | XP_011507445.1 | 6 | 949 | |||
XM_011509144.1 | XP_011507446.1 | 6 | 952 | |||
XM_011509147.2 | XP_011507449.1 | 6 | 939 | |||
XM_017000241.1 | XP_016855730.1 | 6 | 942 | |||
XM_017000242.1 | XP_016855731.1 | 5 | 854 | |||
XM_017000243.1 | XP_016855732.1 | 4 | 1982 | |||
XM_017000244.1 | XP_016855733.1 | 6 | 936 | |||
GenBank mRNA | AK091036.1 | - | 2 | 1155 | ||
AK291995.1 | - | 6 | 811 | |||
AK303696.1 | - | 6 | 658 | |||
BC006274.2 | - | 2 | 155 | |||
BC014067.2 | - | 6 | 722 | 3837777 | ||
BC053603.1 | - | 6 | 885 | 6007375 | ||
L29216.1 | - | 5 | 679 | |||
L29218.1 | - | 6 | 767 |