Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CDH12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001317227.1 | NP_001304156.1 | 6 | 1325 | ||
NM_001317228.1 | NP_001304157.1 | 6 | 1320 | |||
NM_004061.4 | NP_004052.2 | 9 | 1954 | |||
XM_011513927.2 | XP_011512229.1 | 7 | 1231 | |||
XM_017008920.1 | XP_016864409.1 | 8 | 1581 | |||
XM_017008921.1 | XP_016864410.1 | 9 | 1674 | |||
GenBank mRNA | AK092176.1 | - | 4 | 604 | ||
AK295123.1 | - | 6 | 1320 | |||
AK314800.1 | - | 6 | 1325 | |||
BC047608.1 | - | 9 | 1659 | 5751948 | ||
L33477.1 | - | 9 | 1955 | |||
L34057.1 | - | 5 | 960 |