Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CDMO-CRO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other CHN2 SIRNA Assays ›
Gene Symbol:
CHN2
Gene Name:
chimerin 2
Gene Aliases:
ARHGAP3, BCH, CHN2-3, RHOGAP3
Chromosome Location:
Chr.7: 29146547 - 29514335 on Build GRCh38
UniGene ID:
Hs.654611
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
146299
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: CHN2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001039936.2
NP_001035025.1
3
773
NM_001293069.1
NP_001279998.1
10
1044
NM_001293070.1
NP_001279999.1
10
1258
NM_001293071.1
NP_001280000.1
8
1163
NM_001293072.1
NP_001280001.1
9
976
NM_001293073.1
NP_001280002.1
3
638
NM_001293075.1
NP_001280004.1
3
638
NM_001293077.1
NP_001280006.1
3
773
NM_001293079.1
NP_001280008.1
3
773
NM_001293080.1
NP_001280009.1
3
773
NM_001293081.1
NP_001280010.1
3
288
NM_004067.3
NP_004058.1
9
1219
NR_120524.1
2
553
XM_011515105.2
XP_011513407.1
12
1099
XM_011515106.2
XP_011513408.1
11
1060
XM_011515107.2
XP_011513409.1
11
1842
XM_017011721.1
XP_016867210.1
13
1117
XM_017011722.1
XP_016867211.1
12
1860
GenBank mRNA
AK026415.1
-
3
773
AK293959.1
-
10
838
AK294032.1
-
9
976
AK294216.1
-
3
288
AK294271.1
-
10
1045
AK301235.1
-
3
297
AK313021.1
-
9
993
AK316030.1
-
3
297
BC112155.1
-
9
964
8327623
EU732750.1
-
2
154
EU732752.1
-
3
239
EU732753.1
-
3
239
EU732756.1
-
3
374
EU732757.1
-
3
374
EU732758.1
-
3
374
EU732759.1
-
2
92
EU732760.1
-
8
726
EU732762.1
-
6
350
GQ924106.1
-
10
1045
U28926.1
-
9
1016
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
positive regulation of signal transduction
intracellular signal transduction
positive regulation of GTPase activity
regulation of small GTPase mediated signal transduction
SH3/SH2 adaptor activity
GTPase activator activity
protein binding
metal ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-5c9c5c4749-x88h5:80/100.66.78.96:80.
git-commit: d22414ba47b22cc02ffc24003f0e179da61511a6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.21.0-Offline