Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DCTN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135040.2 | NP_001128512.1 | 16 | 2486 | ||
NM_001135041.2 | NP_001128513.1 | 14 | 2209 | |||
NM_001190836.1 | NP_001177765.1 | 16 | 2429 | |||
NM_001190837.1 | NP_001177766.1 | 18 | 2525 | |||
NM_004082.4 | NP_004073.2 | 19 | 2546 | |||
NM_023019.3 | NP_075408.1 | 14 | 2209 | |||
NR_033935.1 | 16 | 2429 | ||||
GenBank mRNA | AB209625.1 | - | 10 | 3045 | ||
AK297286.1 | - | 16 | 2181 | |||
AK314352.1 | - | 16 | 2430 | |||
AK315014.1 | - | 14 | 2001 | |||
BC071583.1 | - | 14 | 2126 | 4817849 | ||
BX640799.1 | - | 19 | 2325 | |||
BX647925.1 | - | 16 | 2190 | |||
BX647972.1 | - | 16 | 2209 | |||
U90445.1 | - | 7 | 1004 | |||
X98801.1 | - | 17 | 2183 |