Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DCTN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135040.2 | NP_001128512.1 | 18, 19 | 2732 | ||
NM_001135041.2 | NP_001128513.1 | 16, 17 | 2455 | |||
NM_001190836.1 | NP_001177765.1 | 18, 19 | 2675 | |||
NM_001190837.1 | NP_001177766.1 | 20, 21 | 2771 | |||
NM_004082.4 | NP_004073.2 | 21, 22 | 2792 | |||
NM_023019.3 | NP_075408.1 | 16, 17 | 2455 | |||
NR_033935.1 | 18, 19 | 2675 | ||||
GenBank mRNA | AB209625.1 | - | 12 | 3453 | ||
AK297286.1 | - | 18, 19 | 2427 | |||
AK314352.1 | - | 18, 19 | 2676 | |||
AK315014.1 | - | 16, 17 | 2247 | |||
BC071583.1 | - | 16, 17 | 2372 | 4817849 | ||
BX640799.1 | - | 21, 22 | 2571 | |||
BX647925.1 | - | 18, 19 | 2436 | |||
BX647972.1 | - | 18, 19 | 2455 | |||
X98801.1 | - | 19, 20 | 2429 |