Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DCTN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135040.2 | NP_001128512.1 | 19, 20 | 2883 | ||
NM_001135041.2 | NP_001128513.1 | 17, 18 | 2606 | |||
NM_001190836.1 | NP_001177765.1 | 19, 20 | 2826 | |||
NM_001190837.1 | NP_001177766.1 | 21, 22 | 2922 | |||
NM_004082.4 | NP_004073.2 | 22, 23 | 2943 | |||
NM_023019.3 | NP_075408.1 | 17, 18 | 2606 | |||
GenBank mRNA | AB209625.1 | - | 12, 13 | 3604 | ||
AK297286.1 | - | 19, 20 | 2578 | |||
AK314352.1 | - | 19, 20 | 2827 | |||
AK315014.1 | - | 17, 18 | 2398 | |||
BC071583.1 | - | 17, 18 | 2523 | 4817849 | ||
BX640799.1 | - | 22, 23 | 2722 | |||
X98801.1 | - | 20, 21 | 2580 |