Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EFNB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004093.3 | NP_004084.1 | 5 | 1895 | ||
XM_017020406.1 | XP_016875895.1 | 5 | 10156 | |||
GenBank mRNA | BC105955.1 | - | 5 | 1840 | 40034079 | |
BC105956.1 | - | 5 | 1840 | 40034078 | ||
BC105957.1 | - | 5 | 1840 | 40034080 | ||
L38734.1 | - | 5 | 1850 | |||
U81262.1 | - | 5 | 1826 |