Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EPRS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004446.2 | NP_004437.2 | 4, 5 | 666 | ||
GenBank mRNA | AK296755.1 | - | 4, 5 | 538 | ||
AK307534.1 | - | 2, 3 | 240 | |||
AY493416.1 | - | 2, 3 | 213 | |||
BC015494.1 | - | 4, 5 | 454 | |||
BC034797.1 | - | 4, 5 | 462 | |||
BC046156.1 | - | 4, 5 | 666 | |||
BC058921.1 | - | 4, 5 | 503 | |||
BC067841.1 | - | 4, 5 | 538 | |||
BC105983.1 | - | 4, 5 | 464 | |||
BC126275.1 | - | 4, 5 | 534 | 8991991 | ||
BC136465.1 | - | 4, 5 | 434 | 9020454 | ||
CR933648.1 | - | 4, 5 | 505 | |||
JQ080075.1 | - | 4, 5 | 396 | |||
X54326.1 | - | 2, 3 | 238 |