Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EIF4EBP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004096.4 | NP_004087.1 | 3 | 1584 | ||
GenBank mRNA | BC005057.1 | - | 3 | 1310 | 2823322 | |
BC050633.1 | - | 3 | 1523 | 5555034 |