Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STOM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270526.1 | NP_001257455.1 | 7 | 888 | ||
NM_004099.5 | NP_004090.4 | 7 | 979 | |||
NM_198194.2 | NP_937837.1 | 3 | 484 | |||
NR_073037.1 | 8 | 1109 | ||||
GenBank mRNA | AK302977.1 | - | 7 | 905 | ||
AK304449.1 | - | 8 | 1053 | |||
AK304518.1 | - | 6 | 620 | |||
AK307999.1 | - | 7 | 922 | |||
BC010703.1 | - | 7 | 901 | 3873494 | ||
BG701899.1 | - | 3 | 437 | |||
BM925356.1 | - | 1 | 61 | |||
M81635.1 | - | 7 | 883 | |||
X60067.1 | - | 7 | 862 |