Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ETFDH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004453.3 | NP_004444.2 | 2, 3 | 523 | ||
GenBank mRNA | AK314803.1 | - | 2, 3 | 500 | ||
BC011890.1 | - | 2, 3 | 325 | 3938660 | ||
BX538129.1 | - | 2, 3 | 294 | |||
BX957055.1 | - | 2, 3 | 325 | |||
DA319656.1 | - | 2, 3 | 499 | |||
EU093082.1 | - | 2, 3 | 219 | |||
S69232.1 | - | 2, 3 | 287 |